Visualization interpretation guidance
Oncomine™ Lung Cell‑Free Total Nucleic Acid Research Assay
Metric | Description |
|---|---|
Copy Number Variation | |
CNV Ratio | Should be interpreted as the fold amplification (gain) as detected by the assay. CNV specific amplicon (MET) coverage levels are compared to non-CNV amplicon coverage. |
P-value | Significance of CNV Ratio measurement based on amplicon coverage variability (MAPD level) and magnitude of the pairwise coverage differences between the CNV and non-CNV amplicons. High coverage variability will result in less significant p-values. |
Fusion detection | |
Nomenclature | Each reported fusion target follows a specific naming convention such that the 5'- and 3'‑genes are reported along with donor and acceptor exon numbers. Lastly, a COSMIC ID or NCBI transcript accession number is added to the end of each target name. For example, EML4-ALK.E13A20.COSF463 identifies the EML4-ALK fusion variant with exon 13 of EML4 fused to exon 20 of ALK. |
Fusion QC genes | Two non-fused process control genes (HMBS and TBP) that have been shown to be consistently detected in cell-free nucleic acid extracts are included in the assay to inform quality of fusion variant calls. |
Analysis detail |
|
MET Exon 14 Skipping Assay | |
Nomenclature |
|
Analysis detail |
|
Oncomine™ Breast cfDNA Research Assay v2
Metric | Description |
|---|---|
Copy Number Variation | |
CNV Ratio | Should be interpreted as the fold amplification (gain) as detected by the assay. CNV specific amplicon (CCND1, ERBB2, FGFR1) coverage levels are compared to non-CNV amplicon coverage. |
P-value | Significance of CNV Ratio measurement based on amplicon coverage variability (MAPD level) and magnitude of the pairwise coverage differences between the CNV and non-CNV amplicons. High coverage variability will result in less significant p-values. |
De novo (non‑hotspot) variant calling in TP53 | |
Analysis detail |
|