Unable to parse header in IGV
Unable to parse header:
Your input file has a malformed header: unknown column name 'CHR'; it does not match a legal column header name.
Possible cause | Recommended action |
|---|---|
|
IGV does not load variants and displays this error message when a VCF file uses CHR instead of CHROM for a column header name. |
To avoid this error, use the CHROM column header name in input VCF files. For example, #CHROM POS ID REF. |