Set IRGV preferences
You can modify the elements that are included in Ion Reporter™ Genomic Viewer (IRGV) analysis visualizations with preferences. To set the preferences, you must first visualize an analysis in the viewer.
  1. Under the Analyses tab, click Overview.
  2. Select one or more analyses in the Analysis Results screen, then click Visualize.
  3. In the IRGV & Generate Report tab, scroll to the bottom of the visualization, then click IRGV/Export & PreferencesShow IRGV preferences to expand the Preferences section.
    You can set the following IRGV preferences.
    Option
    Description
    Default mapd value
    This value is used as default value in the mapd filter.
    Default sort order [1]
    The preferred sort order for Aneuploidy analyses (NR, CID, EID or BIOPSY) for the summary table and tracks. NR sorts by the analysis nr, CID sorts by couple ID, EID sorts by embryo ID and BIOPSY sorts by biopsy days.
    Analysis limit for BAM tracks
    Maximum number of analysis that includes BAM tracks in IRGV (if available). Loading higher number of tracks would result in longer loading time and a crowded view.
    Analysis limit for BED tracks
    Maximum number of analysis that includes BED tracks in IRGV (if available). Loading higher number of tracks would result in longer loading time and a crowded view.
    Limit for coverage data
    By default, the maximum limit of 25000 is used. Decrease this number if you do not want to see the coverage data, or you have problems with the browser, such as freezing.
    Max number tracks in karyo
    By default, 5 tracks are shown. You can set a value of 1 to 10. When you increase the number of tracks, more space is used in the Karyo view.
    Max Ploidy in Whole Genome View
    Maximum value of the Y-axis (ploidy) in the Whole Genome View graphs.
    Display Fixed Ploidy Lines
    You can show fixed ploidy lines at copy number=1 and copy number=3 across the Whole Genome View.
    • true—Select this default setting to show fixed ploidy lines at CN=1 and CN=3 across the Whole Genome View.
    • false—Select this setting to show ploidy lines that are not fixed.
    Whole Genome View Height
    You can double the height of the Y-axis when you view whole genome data in IRGV. The Enlarged view is useful to review mosaic data, because more data are shown on a single plot and more space is available between data points, so that the mosaic data more pronounced.
    • Compressed—Select this default setting for IRGV images that are 300 pixels high.
    • Expanded—Select this setting for IRGV images that are 600 pixels high.
    1 This preference can be set only when multiple analyses are selected.
  4. For each value that you want to change in the Preferences section, click Edit next to the value that you want to change, then enter a new value into the Edit Preferences dialog box, then click OK.
  5. When the edits are complete, click Save Preferences.
    The preferences are reset to the default settings if you click Reset Preferences.
    The selected preferences are applied to all analyses that you visualize immediately after you save the changes.