Example RNA Exon Tile Fusion Imbalance read count plot for a single analysis
RNA Exon Tile Fusion Imbalance plots provide a visual representation of the RNA fusion imbalance analysis results for the detection of novel fusions in a partner agnostic manner. The Normalized Read Depth by Gene plot shows the mean read counts of each gene that is captured on the chip for the selected sample. For each gene, the read counts are normalized to the number of amplicons.