3'/5' Imbalance scores for assay calls
Panels that include 3'/5' amplicons generate 3'5 imbalance values. The imbalance values report the difference in expression between the 5' assay and the 3' assay of each driver gene. Samples that do not contain a fusion are expected to have similar expression of the 5' assay compared to the 3' assay of the driver gene. Samples that contain a fusion are often expected to have elevated expression of the 3' assay compared to the 5' assay.
The 3'/5' Imbalance values provide a measurement of the strength of fusion calls that involve ALK, RET, or ROS1 driver genes, whether included or not included in the Ion AmpliSeq RNA Fusion Lung Cancer Research Panel.
The 3'/5' Imbalance score is therefore included for two alternate purposes:
  • To confirm presence of a fusion from the ALK, RET, or ROS1 driver genes included in the panel, as shown in this figure:
  • If none of the fusions targeted by the panel is detected, to provide evidence of a fusion other than those targeted by the panel but still including ALK, RET, or ROS1 driver genes.
The following approaches interpret the 3'/5' Imbalance values for each gene and are reliable only under the conditions that are described in the following sections:
Most samples that are tested for the 3'/5' Imbalance assay have been lung tumor tissue. Threshold settings for other sample types can be sensitive to varying expression of fusion gene and expression control genes.