Sensitivity
The analysis workflow is very sensitive for fusion detection and can possibly pick up noise if there are problems with, for instance, the sample or library preparation.
Check for High 5' expression and also check for the minimum number of reads before assuming that a fusion call of 'Present' is confirmed.