View 3'/5' Imbalance scores
You can detect novel fusions of select driver genes in Ion Reporter™ Software with 3'/5' Imbalance scores that are shown in analysis results.
- In the Analyses tab, click Overview.
-
Click an analysis name.
The Analysis Results screen opens to the Fusions table.