Example Normalized Read Depth by Gene RNA Exon Tile Fusion Imbalance plot for multiple analyses
RNA Exon Tile Fusion Imbalance plots provide a visual representation of the RNA fusion imbalance analysis results for the detection of novel fusions in a partner agnostic manner. The Normalized Read Depth by Gene plot shows the mean read counts of each gene that is captured on the chip. For each gene, the read counts are normalized to the number of amplicons. For multi-analysis visualization, the data are displayed side by side. The number of bars corresponds to the number of analyses.