Example of an analysis with no fusions called
This image shows an example of the Analysis Results table for a run in which no fusions are called.
Note: When you sort on the Gene column, the software sorts on both of the genes involved in the fusion. As a result, the lexicographically smaller gene is used for an ascending sort, while the lexicographically larger gene is used for a descending sort.