Analysis workflow revision autonumbering
Ion Reporter™ Software displays revision numbers of analysis workflows, and automatically increases the revision numbers when you copy or edit an analysis workflow.
To view analysis workflow version numbers, click , then select the desired analysis workflow. The analysis workflow version number, for example w2.2, appears in two places.
-
In the Workflow Name column at the end of the analysis workflow name.
-
In the Details section at the end of the analysis workflow name.
The version number increases if you copy a predefined analysis workflow, or copy and edit a user-defined analysis workflow and use the name of the previous analysis workflow as the name for the new analysis workflow.
You can copy predefined analysis workflows and user-defined analysis workflows from the current version of the software or from previous versions of the software.