About analysis workflows
Analysis workflows in Ion Reporter™ Software are sets of instructions that determine how analysis results are produced.
The predefined analysis workflows include settings for common genetic research analysis applications. You can use a predefined analysis workflow as is, or you can copy a predefined analysis workflow, customize its settings, then save and reuse it in the future. After a predefined analysis workflow is copied and saved, it is known as a user-defined analysis workflow. You can also copy and edit, or edit a user-defined analysis workflow.
When an analysis workflow that contains samples is launched, Ion Reporter™ Software generates an analysis. Each analysis contains analysis results in various formats, including tables and visualizations in the software, and data files that can be downloaded.
In Torrent Suite™ Software, you can create a Planned Run that transfers data automatically to the appropriate Ion Reporter™ Server or to an Ion Reporter™ Software on Connect account, and use any of the available analysis workflows to create an analysis.
All analysis workflows that are available to the Ion Reporter™ Software organization are listed under the Workflows tab.