Get user details
getUserDetails
URL
/api/v1/getUserDetails
Description
Used by IonReporterUploader plugin to get user details based on username and password. Returns the user details.
Method
POST
Request query parameters
  • userName
    String, Mandatory.
    Valid username.
  • password
    String, Mandatory.
    Valid password.
Example request
https://user1.XYZ.com/api/v1/getUserDetails
Request headers
Content-Type: application/x-www-form-urlencoded
Returns
{
  "tokendate": "2013-12-08 15:39:37.0",
  "firstname": "Ion",
  "eulaAccepted": "true",
  "userStatus": "ENABLED",
  "orgname": "IR Org",
  "exporttokendate": "2015-11-05 03:25:12.0",
  "exporttoken": "49bbe49a8307ba61ef28ee9a9122a98347576488c975c857b7e1c82978f3818c",
  "lastname": "User",
  "token": "wVcoTeYGfKxItiaWo2lngsV/r0jukG2pLKbZBkAFnlPbjKfPTXLbIhPb47YA9u78"
}
Response fields
Status code
Response message
200
Successful response
400
Bad request
404
Not found
401
User is not authenticated
500
Error occurred
Curl command
curl -X POST -k https://<server name>/api/v1/getUserDetails -H "Content-Type:application/x-www-form-urlencoded" -d "userName=ion.reporter@lifetech
.com&password=IonPass123"