Download QC and final reports
To retrieve one Quality Control (QC) report in JSON format, enter the following.
curl --request GET -k -H "Authorization:ZDMxNDUwZGYyNjM4NGRlZjY0NjAyNTc1YTE1ZjU2MzFkNjVmYzQxYjJhOTQwNTYyYWM3OTJmZDY2YTU5NGQwNA" -O "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/new_CFTR/new_CFTR_20151106002127527/new_CFTR_1446798039543.zip"
The output in JSON format resembles this:
[
{
"reports": {
"qc": {
"link": "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/download/pdf/335a1f5e-19f1-4ff4-b80c-af620f1dfe1f/ff80818150dbc2430150dbe2d9650072_QC.pdf"
},
"final": {}
},
"report_published": "",
"workflow": "AmpliSeq CFTR single sample",
"data_links": {
"filtered_variants": "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/new_CFTR/new_CFTR_20151106002127527/new_CFTR_1446798039543.zip",
"unfiltered_variants": "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/new_CFTR/new_CFTR_20151106002127527/new_CFTR_1446798039543.zip"
},
"variants_saved": "",
"samples": {
"PROBAND": "new_CFTR"
},
"shared_with": [],
"stage": "Review Variants",
"flagged": false,
"name": "new_CFTR_1446798039543",
"id": "ff80818150dbc2430150dbe2d9650072",
"ion_reporter_version": "5.0",
"started_by": "Ion User",
"start_date": "November 06, 2015",
"status": "SUCCESSFUL"
}
]
To retrieve all of the QC reports in JSON format, enter:
curl --location "https://<server name>/api/v1/qcreport?format=json" \
--header "Authorization: OTAzNzY1ZjMzY2U4YmE5NzhmN2RhYTFkYTJmMzI2NTE1OGI2YWMzMjRkMGM5\OTI1MDkzNWMxMjFiOWRjYWI4MA" \
--header "Content-Type: application/x-www-form-urlencoded"
To retrieve one final report in JSON format, enter:
curl --location "https://<server name>/api/v1/finalreport?format=json&name=Demo%20Aneuploidy_c23157_2025-05-1010-03-46-508" \
--header "Authorization: OTAzNzY1ZjMzY2U4YmE5NzhmN2RhYTFkYTJmMzI2NTE1OGI2YWMzMjRkMGM5OTI1MDkzNWMxMjFiOWRjYWI4MA" \
--header "Content-Type: application/x-www-form-urlencoded"
To retrieve all of the final reports in JSON format, enter:
curl --location "https://<server name>/api/v1/analysis?format=json&name=Demo%20AmpliSeq%20CCP%20tumor_prod" \
--header "Authorization: OTAzNzY1ZjMzY2U4YmE5NzhmN2RhYTFkYTJmMzI2NTE1OGI2YWMzMjRkMGM5OTI1MDkzNWMxMjFiOWRjYWI4MA" \
--header 'Content-Type: application/x-www-form-urlencoded' 