Get QC report
qcreport
URL
/api/v1/qcreport
Description
Gets the Quality Control (QC) Report. Returns a JSON object that contains download information.
Method
GET
Request query parameters
  • name
    String, Mandatory.
    Name of sample or analysis.
  • format.
    String. Optional.
    Response type, current version supports only json.
  • type
    String, Optional.
    Valid values are "sample" or "analysis".
    If the type parameter is not provided, "analysis" is used as the default value.
  • id
    String. Optional.
    ID of an analysis.
  • start_date.
    String, Optional.
    Parameter to filter the results by date boundary. Date format should be in yyyy-MM-dd format.
  • end_date
    String, Optional.
    Parameter to filter the results by date boundary. Date format should be in yyyy-MM-dd. format.
  • view
    String,Optional.
    Valid values are "summary" or "" .
Example request
https://user1.XYZ.com/api/v1/qcreport?fomat=json&type=analysis&
name=Demo AmpliSeq Exome VCF_1460407585676&start_date=2016-01-30&end_date=2016-04-12
Request headers
Content-Type: application/x-www-form-urlencoded
Authorization:API Token
Returns
[
  {
    "stage": "VARIANT_REVIEW",
    "flagged": false,
    "name": "Demo AmpliSeq Exome VCF_1460407585676",
    "id": "ff80818153ed40a401540716785a081a",
    "qc_metrics": {
      "Demo AmpliSeq Exome VCF": {
        "Number of Heterozygous SNVs": " 32451",
        "CNV/Total": " 0.000",
        "Number of Homozygous INDELs": " 1089",
        "Ti/Tv Ratio (SNPs)": " 2.308",
        "MAPD": "N/A",
        "LongDels/Total": " 0.000",
        "Number of Heterozygous INDELs": " 2113",
        "Heterozygotes/Homozygotes": " 1.737",
        "Indels/kb": " 0.001",
        "Number of Heterozygous MNVs": " 0",
        "Number of LongDels": " 0",
        "Number of Homozygous MNVs": " 0",
        "Indels/Total": " 0.059",
        "SNPs/kb": " 0.017",
        "Number of Homozygous SNVs": " 18814",
        "Number of CNVs": " 0",
        "dbSNP concordance": " 0.934"
      }
    },
    "started_by": "Ion User",
    "ion_reporter_version": "5.22",
    "samples": {
      "PROBAND": "Demo AmpliSeq Exome VCF"
    },
    "qc_report_pdf": "http://user1.XYZ.com/api/v1/download?type=pdf&
                filePath=/data/IR/data/IR_Org/download/pdf/
                890e4268-46dc-442b-a4ed-aa87bb28f71a/
                ff80818153ed40a401540716785a081a_QC.pdf",
    "start_date": "2016-04-11 13:49:53.0",
    "status": "SUCCESSFUL",
    "qc_package": "http://user1.XYZ.com/api/v1/download?
                filePath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/
                Demo_AmpliSeq_Exome_VCF/Demo_AmpliSeq_Exome_VCF_20160411134952720/
                analysis_downloads/QC_Bundle/
                qc-report-Demo_AmpliSeq_Exome_VCF_1460407585676_2025-04-12 04-33.zip"
  }
]
Response fields
Status code
Response message
200
Successful response
400
Bad request
404
Not found
401
User is not authenticated
500
Error occurred
Curl command
curl -v -k -X GET "https://<server name>/api/v1/qcreport?format=json&name=BRCA_Plus_RR_jul29&type=analysis&start_date=2016-07-25&end_date=2016-07-31" -H "Content-Type:application/x-www-form-urlencoded" -H "Authorization:NDliYmU0OWE4MzA3YmE2MWVmMjhlZTlhOTEyMmE5ODM0NzU3NjQ4OGM5NzVjODU3YjdlMWM4Mjk3OGYzODE4Yw"