Query by sample name API call You can query by sample name using this API call: /api/v1/analysis?format=json&name=[samplename]&type=sample This call returns a list of analysis names performed on the sample, ordered by start date.