Send an analysis to the Report role
You can send an analysis to someone in the organization who has the Report role in Ion Reporter™ Software. A person with the Report role can then generate a report.
- In the Analyses tab, select Overview.
- Click an analysis link to open the Analysis Results screen.
-
Click .
Note: The Send to Report Role button activates the same function.
- In the Send to Report Role window, click Send.
The analysis results are sent to the person in the organization who has the Report role. The person with the Report role can find the results on the Analyses Overview screen. The results are at the Pending Report stage.