Share analyses data
You can share analysis data from Ion Reporter™ Software with anyone, whether or not they are users of the software.
Variants cannot be flagged when an analysis report has been shared by a user in another organization. The flag column on the Analysis tab and the option to flag multiple variants are not available for shared analysis reports.
When a share is in progress, the analysis and the samples associated with it cannot be edited, archived, or deleted.
  1. In the Analyses tab, select the analysis that contains the data that you want to share, then in the analysis Details click ActionsShare .
  2. In the Share Analysis dialog box, enter the email address of the person with whom you want to share data, then click Share.
    A message about the status of the share opens briefly. When a share is in progress, you see this icon: . You can continue with other tasks in Ion Reporter™ Software while the share is in progress. When the sharing is complete, you and the recipients of the shared data see this icon beside the report that contains shared data: .
  3. Click the Home tab, then click Notifications to view notifications about the share.