Set default number of rows displayed in tables
You can set a default number of rows displayed in all tables in the Ion Reporter™ Software.
- In the upper right corner of the screen, click
-
In the account Preference dialog box, select the number of rows to display in all tables from the Default rows per page dropdown menu, then click Save.
The new settings are applied to all tables within the Ion Reporter™ Software.
- (Optional) To override the default setting for a specific table, navigate to the table, then select the number of rows you want to display from the items per page dropdown menu located below the table.