Ion Reporter Server
Always access Ion Reporter Server through HTTPS (not HTTP).
The URL depends on your server configuration, and is named https://ion-reporter-server-ip-address, where ion-reporter-server-ip-address is the IP address or hostname of your Ion Reporter Server.