User roles and permissions
In Ion Reporter™ Software, access is granted to specific functions based on roles that are assigned to users. Typically, an Ion Reporter™ Software organization has different team members performing different functions. Therefore, each user role within Ion Reporter™ Software has access to different areas of the software.
|
User role |
Access |
|---|---|
|
Import |
The Import user is the person in the Ion Reporter™ Software organization who can import and define samples. Specific access includes the following tasks.
|
|
Analyze |
The Analyze user is the person in the Ion Reporter™ Software organization who can create analysis workflows and launch analyses. Specific access includes the following tasks.
|
|
Report |
The Report user is the person in the Ion Reporter™ Software organization who can generate reports. Specific access includes the following tasks.
|
|
Admin |
The Admin user is the person in the Ion Reporter™ Software organization who can manage user access. Specific access includes the following tasks.
|