Ion Reporter™ Software dashboard
You can open the dashboard in Ion Reporter™ Software, then navigate to various screens, view important information about the software, and access support resources. Click the Home tab, then click Dashboard.
-
Quick links pane
-
Announcements pane
-
Support pane
The Quick links pane includes:
-
Define sample—Opens the Define Samples screen, where you can upload and define samples.
-
View samples—Opens the Samples screen, where you can view, search, sort, or filter samples.
-
Create workflow—Opens the Create Workflow wizard, where you can create a new analysis workflow.
-
View workflows—Opens the Workflows screen, where you can view, search, sort, or filter analysis workflows.
-
Launch analysis—Opens the Launch Analysis wizard, where you can select an analysis workflow, add samples and plugins, and launch an analysis.
-
View analyses—Opens the Analyses screen, where you can view, search, sort, or filter analyses.
The Announcements pane includes a link to the latest Ion Reporter™ Software Release Notes and information on upcoming events.
The Have questions? pane includes:
-
Help section link—Opens the software help menu.
-
Send feedback link—Opens a help topic about how to send feedback regarding your experience with Ion Reporter™ Software.
-
Send an email to ionreporter@lifetech.com—Opens Microsoft™ Outlook and creates a new email addressed to Ion Reporter™ Software support.
-
Contact Web Support—Opens the Services & Support web page.
The What's new in Ion Reporter pane describes new features.
Note: Only the tabs and quick links corresponding to your user role are displayed. For more information, see User roles and permissions.