Sign in to Ion Reporter™ Software on Ion Reporter™ Server
Before you can sign in to Ion Reporter™ Software on Ion Reporter™ Server, you must have a user account on the server. An administrator-level user on the server can create an account for you. To sign in to the software on the server:
  1. In a browser on the same network as the Ion Reporter™ Server, enter the server URL: https://ion-reporter-server-ip-address, where ion-reporter-server-ip-address is the IP address or host name of the server.
  2. Enter your user name and password, then click Sign In.