Create an organization on Ion Reporter™ Server
The first user on Ion Reporter™ Server can create an organization. An organization is more specific than an institute name. It defines a group of users who work together and share samples, data, analyses, and results.
When you create an organization, you automatically are the first administrator-level user of that organization. You can then begin to enter or import data into the software and invite other Ion Reporter™ Server users to the organization to share data and results.
You can belong to only one organization at a time. If you are already a member of an organization on Ion Reporter™ Server, you will not be able to create a new organization until you are removed as a user from your current organization.
  1. On the software sign-in screen click, click Register new account.
  2. In the Request Account dialog box, enter the following information: organization name, organization address, your first and last name, email, phone number.
    If the Ion Reporter™ Server user is connected to the Internet, you must select the checkbox in the dialog box.
  3. Click Submit.
    A new organization is created and you become the organization administrator. You will receive an email with a link to set the password.
  4. Click the link in the email and set the password.
  5. After the password is set, you can sign in using the new password. The first time you sign in, you must accept the Terms and Conditions.
You can now begin using the new Ion Reporter™ Server organization.