Codon differences in chromosome M
Chromosome M has its own genome that is different in 4 codons from the other chromosomes.
Ion Reporter™ Software uses the Homo sapiens mitochondrion, complete genome NCBI Reference Sequence: NC_012920.1.
The new rCRS chrM assembly has 16569 bases. The previous version had 16571 bases.
The 2 coding tables, one for standard chromosomes and one for chromosome M, are listed below. These tables are from the following NCBI site:
The Standard Code (transl_table=1) for chromosomes 1-22, X and Y (NUCLEAR)
By default all transl_table in GenBank flat files are equal to id 1, and this is not shown. When transl_table is not equal to id 1, it is shown as a qualifier on the CDS feature.
Note: In these tables, an asterisk represents a STOP codon.
TTT F Phe TCT S Ser TAT Y Tyr TGT C Cys
TTC F Phe TCC S Ser TAC Y Tyr TGC C Cys
TTA L Leu TCA S Ser TAA * Ter TGA * Ter
TTG L Leu i TCG S Ser TAG * Ter TGG W Trp CTT L Leu CCT P Pro CAT H His CGT R Arg
CTC L Leu CCC P Pro CAC H His CGC R Arg
CTA L Leu CCA P Pro CAA Q Gln CGA R Arg
CTG L Leu i CCG P Pro CAG Q Gln CGG R Arg ATT I Ile ACT T Thr AAT N Asn AGT S Ser
ATC I Ile ACC T Thr AAC N Asn AGC S Ser
ATA I Ile ACA T Thr AAA K Lys AGA R Arg
ATG M Met i ACG T Thr AAG K Lys AGG R Arg GTT V Val GCT A Ala GAT D Asp GGT G Gly
GTC V Val GCC A Ala GAC D Asp GGC G Gly
GTA V Val GCA A Ala GAA E Glu GGA G Gly
GTG V Val GCG A Ala GAG E Glu GGG G Gly The Vertebrate Mitochondrial Code (transl_table=2) for chromosome M.
TTT F Phe TCT S Ser TAT Y Tyr TGT C Cys
TTC F Phe TCC S Ser TAC Y Tyr TGC C Cys
TTA L Leu TCA S Ser TAA * Ter TGA W Trp
TTG L Leu TCG S Ser TAG * Ter TGG W Trp CTT L Leu CCT P Pro CAT H His CGT R Arg
CTC L Leu CCC P Pro CAC H His CGC R Arg
CTA L Leu CCA P Pro CAA Q Gln CGA R Arg
CTG L Leu CCG P Pro CAG Q Gln CGG R Arg ATT I Ile i ACT T Thr AAT N Asn AGT S Ser
ATC I Ile i ACC T Thr AAC N Asn AGC S Ser
ATA M Met i ACA T Thr AAA K Lys AGA * Ter
ATG M Met i ACG T Thr AAG K Lys AGG * Ter GTT V Val GCT A Ala GAT D Asp GGT G Gly
GTC V Val GCC A Ala GAC D Asp GGC G Gly
GTA V Val GCA A Ala GAA E Glu GGA G Gly
GTG V Val i GCG A Ala GAG E Glu GGG G Gly The differences from the Standard Code are:
Code 2 | Standard | |
|---|---|---|
AGA | Ter * | Arg R |
AGG | Ter * | Arg R |
AUA | Met M | Ile I |
UGA | Trp W | Ter * |
Background information
The UCSC website has this note about chrM:
“Note on chrM:
Since the release of the UCSC hg19 assembly, the Homo sapiens mitochondrion sequence (represented as "chrM" in the Genome Browser) has been replaced in GenBank with the record NC_012920. We have not replaced the original sequence, NC_001807, in the hg19 Genome Browser. We plan to use the Revised Cambridge Reference Sequence (rCRS) in the next human assembly release.”
The IGSR: The International Genome Sample Resource has this entry:
Which reference assembly do you use?
The reference assembly the 1000 genomes project has mapped sequence data that has changed over the course of the project.
For the pilot phase we mapped data to NCBI36. A copy of our reference FASTA file can be found on the ftp site.
For the phase1 and phase 3 analysis we mapped to GRCh37. Our FASTA file which can be found here called human_g1k_v37.fasta.gz, it contains the autosomes, X, Y and MT but no haplotype sequence or EBV.
Other links related to chrM:
Original sequence, NC_001807:
http://www.ncbi.nlm.nih.gov/nuccore/NC_001807.4?report=genbank
The updated Mitochondrial sequence, NC_012920 can be found here: