SIFT score
A SIFT score predicts whether an amino acid substitution affects protein function.
The SIFT score ranges from 0.0 (deleterious) to 1.0 (tolerated). The score can be interpreted as follows:
  • 0.0 to 0.05 -- Variants with scores in this range are considered deleterious. Variants with scores closer to 0.0 are more confidently predicted to be deleterious.
  • 0.05 to 1.0-- Variants with scores in this range are predicted to be tolerated (benign). Variants with scores very close to 1.0 are more confidently predicted to be tolerated.