Import a custom annotation source
You can use custom annotation source files in Ion Reporter™ Software. If you have annotation data that you want to use, you need to first create a new annotation set for the custom annotation source. Then, you can import the files into the new annotation set.
There are 4 types of annotation sources that you can import into Ion Reporter™ Software. The types of annotation sources and the file formats required for each annotation source are as follows.
Note: Only one version of an annotation source can be used in an annotation set.
|
Annotation source type |
Description |
File format for import |
|---|---|---|
|
Genomic region |
A set of regions |
BED |
|
Gene set |
A set of genes |
TXT |
|
Preferred transcript set |
A set of transcripts |
TXT |
|
VariantDB |
A set of variants |
VCF |
- In the Workflows tab, click Presets.
- Click .
- Enter a descriptive Name for the custom annotation set.
- (Optional) Enter a description for the custom annotation set.
-
Select a custom annotation type from the dropdown list:
-
Genomic Region (Custom)
-
Gene Set (Custom)
-
Preferred transcript set (Custom)
-
VariantDB (Custom)
-
- Click the Create New tab.
-
Enter a descriptive Name and Version for the custom annotation source.
IMPORTANT! When naming custom annotation sources and annotation versions:
-
Do not use capitalized AND, NOT, or OR. You can use lower case and, not, or or.
-
You cannot use two consecutive underscores (__).
You cannot save the object using these characters.
-
- (Optional) Enter a description for the custom annotation source.
- Click Select File, then find and select the custom annotation source file, then click Open.
- Click Save.
- The new custom annotation source is added to the custom annotation set.