Chromosome M
Ion Reporter™ Software uses the Cambridge Reference Sequence (rCRS) for chromosome M with the GenBank accession number NC_012920. Ion Reporter™ Software uses the UCSC curated RefSeq genes which currently do not contain chrM genes. To view gene annotation on chrM use the ENSEMBL gene model, which does have chrM genes.
More information is available at: http://genome.ucsc.edu/cgi-bin/hgGateway?org=human&db=hg19.