Ambiguity codes for three positions on chromosome 3
Three positions on chromosome 3 are marked with 'N' in the UCSC version of the genome. These positions have IUPAC ambiguity codes in our version.
|
Position |
IUPAC ambiguity code in Ion reference |
Hard masked character in UCSC hg19 |
|---|---|---|
|
60830534 |
M |
N |
|
60830763 |
R |
N |
|
60830764 |
R |
N |