Targeted fusion detection method
In the targeted fusion detection method, panel primers are designed to target specific exon-exon junctions of fusions where the driver gene, the partner gene, and the breakpoint between the driver and the partner gene are known. The sequencing reads are mapped to a reference file that contains only the known gene fusions.