Demonstration samples
You can use the predefined analysis workflows with the demonstration samples that are included in Ion Reporter™ Software. You can use these samples to practice the steps to launch analyses and view analysis results.
|
Analysis workflow to use with the demonstration sample |
Demonstration sample name |
Detection type |
|---|---|---|
|
Ion AmpliSeq™ CCP single sample |
Demo Ion AmpliSeq™ CCP tumor |
Somatic |
|
Ion AmpliSeq™ CCP tumor-normal pair |
Demo Ion AmpliSeq™ CCP normal and Demo Ion AmpliSeq™ CCP tumor |
Somatic |
|
Ion AmpliSeq™ CCP paired sample |
Demo Ion AmpliSeq™ CCP CNV control and Demo Ion AmpliSeq™ CCP CNV case, or Demo AmpliSeq CCP normal and Demo Ion AmpliSeq™ CCP tumor |
Somatic |
|
Ion AmpliSeq™ Exome paired sample |
Demo Ion AmpliSeq™ Exome CNV control and Demo Ion AmpliSeq™ Exome CNV case |
Germline |
|
Ion AmpliSeq™ Exome single sample (Germline) |
Demo Ion AmpliSeq™ Exome CNV case |
Germline |
|
Ion AmpliSeq™ Exome single sample (Somatic) |
Demo Ion AmpliSeq™ Exome CNV case |
Somatic |
|
Ion AmpliSeq™ IDP single sample |
Demo Ion AmpliSeq™ IDP Daughter |
Germline |
|
Ion AmpliSeq™ IDP trio |
Demo Ion AmpliSeq™ IDP Daughter, Demo Ion AmpliSeq™ IDP Father, and Demo Ion AmpliSeq™ IDP Mother |
Germline |
|
Ion AmpliSeq™ Colon Lung v2 with RNA Lung Fusion single sample |
Demo Ion AmpliSeq™ RNA Lung Fusion |
Somatic |
|
Ion AmpliSeq™ RNA Lung Fusion single sample |
Demo Ion AmpliSeq™ RNA Lung Fusion |
Fusions and Expressions Control |
|
Aneuploidy |
Demo Aneuploidy |
CNV only |
|
Metagenomics 16S w1.1 |
Demo Metagenomics Mock Community |
Bacterial Identification |
|
Annotate variants single sample |
Demo Ion AmpliSeq™ Exome VCF |
Variant Annotation only |