Manually launch an analysis
  1. Launch the analysis.
    From the…
    Directions
    Home tab
    1. In the Dashboard screen, click Launch analysis.
    2. In the Launch Analysis screen, select a Research Application.
    3. In the Research Category list, select a row with an analysis workflow name.
    4. Click Next.
    Analyses tab
    1. Click Launch Analysis, then select Manual from the list.
    2. In the Launch Analysis screen, select a Research Application.
    3. Click in the analysis workflow row, then click Next.
    Workflows tab
    1. Select a Research Application.
    2. Click in the analysis workflow row, then in the Details pane select Launch Analysis from the Actions list.
  2. Search by any unique identifier you used to label your samples during setup. Select one or more of the samples to include in the analysis, then click Next.
  3. (Optional) Select plugins to run with your analysis.
    1. Click (Actions) adjacent to the selected plugin.
    2. Configure the plugin parameters as necessary, click Submit, then click Close.
  4. Click Next.
  5. Enter an Analysis Name and an optional Description, then click Launch Analysis.
The Ion Reporter™ Software performs the analysis and the results appear on the Analyses page.