Delete a user account on Ion Reporter™ Server
An administrator can delete an Ion Reporter™ Software user account . When you delete a user account, the email address for that user can be reused for a different user account.
  1. Click the Admin tab.
  2. In the Users screen, select the user account in the table (without clicking the hyperlink for the account), then in User Details click Actions Delete.
  3. If you are certain that you want to delete the user account, click Yes to confirm this deletion.
The user account is deleted.