Add columns to the Analyses table
You can add columns to the Analyses table that you view in Ion Reporter™ Software. Columns in the Analyses table show values for information about the analysis. The information can be associated with samples that are used in the analysis, with the analysis workflow, or with the analysis itself. Sample attributes can be inherited from Torrent Suite™ Software and include information such as chip type, sequencing run details, and user-defined attributes.
  1. In the Analyses tab, click Overview.
  2. Click PreferencesSelect Columns.
  3. In the Select Columns dialog box, then select the sample attributes that you want to add to the Samples table, or deselect the sample attributes that you want to remove from the table.
The selected attributes are shown the Analyses table until you close the table. To save the table with the attributes that you added, create a preference. For more information, see Create an Analyses table preference.