Restore default settings for the Analyses table
You can restore default table column headings in the Analyses table in Ion Reporter™ Software.
  1. In the Analyses tab, click Overview.
  2. Click PreferencesRestore Defaults.
The default table settings are restored.