Change the order and width of columns in the Analyses table
Default columns are included for all analyses. You can sort the Analyses table to more easily find analyses of interest, either by the date that the analysis was run, or by sample, workflow, or analysis attributes. You can also add other available columns to customize the table.
You can change the order and width of the columns in the Analyses table in Ion Reporter™ Software.
  1. In the Analyses tab, select Overview.
  2. Click any column heading that you want to move, then drag and drop each column to the position that you prefer. Change the width of each column, as desired.
    The column order and width remain as you have arranged them until you sign out of the software.
  3. (Optional) To use the new column order in the future, create a table preference. Save the order of the columns as a table preference.
    1. Click PreferencesSave Table Preference As.
    2. Enter a name for the table preference, then click Save.
    The Analyses table preference is saved.