Download IonReporterUploader command-line utility
This procedure explains how to download and extract the IonReporterUploader command-line utility. The procedure can vary, based on the operating system of the target computer. In general, decompress the ZIP file on the target machine, then copy the folder IonReporterUploader-cli to a convenient location.
The IonReporterUploader command-line utility is an alternative to the IonReporterUploader plugin that is included with Torrent Suite™ Software.
  1. To access the utility, go to http://iru.ionreporter.thermofisher.com/.
  2. Click the file name IonReporterUploader-cli.zip, then download the file to the target computer.
  3. Extract the downloaded IonReporterUploader-cli.zip file, then copy the IonReporterUploader-cli folder to a convenient location on the target computer.