QC package coverage columns
Quality Control (QC) statistics files report coverage distribution. The QC files that contains the statistics can be downloaded. QC report settings can be customized when you create a custom analysis workflow or edit an analysis workflow.
QC package coverage distribution is calculated for each
region and reported in the No, Low, Medium, and High columns in the *_coverage_stastics.txt files.
|
Coverage |
Definition |
|---|---|
|
No |
0 to 0 coverage range |
|
Low |
1 to Maximum Low Coverage |
|
Medium |
Max Low Coverage +1 to Max Medium Coverage |
|
High |
Maximum Medium Coverage +1 to Maximum Coverage |