Create a sample CSV file to define samples
You can define multiple samples in Ion Reporter™ Software, or update existing samples and edit sample attributes with a CSV file. If you do not yet have a sample file, you can create a new file from a template that is available in Ion Reporter™ Software.
  1. In the Samples tab, click Overview.
  2. Click Define SamplesImport from CSV.
  3. In the Samples step, click download example.
    The SampleDefinition.csv file is downloaded.
  4. Open the SampleDefinition.csv file, then enter the sample information. Include the following information:
    • Relative path to the sample file
      You need to include a relative path to the sample BAM file from the root folder for the Ion Reporter™ Software organization of the user. For example, if the organization of the user is: /data/IR/data/OrganizationName/data/IRU_Uploads/example.bam
      Include the following relative path:
      /data/IRU_Uploads/example.bam
    • Sample name
      Sample names can be any combination of alphanumeric characters plus spaces, periods (.), hyphens (-), and underscores (_). There is a 255-character limit.
    • Sample attributes, such as gender.
  5. Save the file.
After you have entered all the relevant sample information into the CSV file, use the CSV file to define the samples. For more information, see Define samples with a CSV file.