Edit a sample
When you edit an existing Ion Reporter™ Software sample, you can add or remove BAM data files, and add, remove, or change sample attributes.
You cannot edit the preinstalled demonstration samples that are provided with Ion Reporter™ Software. For more information, see Demonstration samples.
You cannot edit locked samples. For more information, see Lock a sample.
- In the Samples tab, click Overview.
- In the Samples table, select the sample that you want to edit, then click Edit.
-
Add the BAM data files from the table to the sample.
-
Click Next to move to the Attributes step in the workflow bar, then edit the attributes that are associated with the sample.
Option
Description
Set the value of a preexisting attribute.
-
Click the field in the column of the attribute heading.
-
Enter a value or select a value from a list.
Add an attribute and set its value.
-
Click Add Attribute and select an attribute from the list.
-
Click the field in the column of the attribute heading.
-
Enter a value or select a value from a list.
You can create a sample preset that can be used for future analyses. For more information, see Create a user-defined sample attribute.
-
- Click Next.
- In the Review step, click Save.
The sample or samples are now ready for analysis. For more information, see Launch an analysis.