Define a sample manually
You can define a new sample manually from imported data files or existing samples.
BAM files from multiple sequencing runs can be combined into a single sample for analysis. For example, you can combine BAM files if you are sequencing the same sample multiple times, and you want to combine those multiple samples into a single sample for greater read depth. Or, you can combine samples from different sequencing runs for sample comparisons. In Ion Reporter Software, you can also define a sample from a single VCF file.
Before you manually define a sample, upload BAM or VCF data files into Ion Reporter Software, or use the demonstration data files available in Ion Reporter Software. For more information, see Upload a BAM file to create a sample or samples, Define samples with a CSV file, and Demonstration samples.