Download analysis information
This example uses curl commands to download information about a specific analysis. The output of the first curl command gives us URLs to use in later curl requests.
This curl command requests analysis information for an analysis named IR50_Rc10_Ane_BC18_DeganCases_1014_np:
curl --request GET -k -H "Authorization:ZTRhNWYyNjkzZjhlYjMxMTdmM2FkZGM1ZmQxYmRkNzU2ZTIxODk5OGRjZWM5ZDY2MjMzMTBiOGIwZWE5NmNlNg" "https://<server name>/api/v1/analysis?format=json&name=IR50_Rc10_Ane_BC18_DeganCases_1014_np"
The output is in JSON format:
[
{
"reports": {
"qc": {
"link": "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/download/pdf/98ccb759-010f-4c92-acba-2da2d8aa9f4d/ff8081815067ca42015068f99193000d_QC.pdf"
},
"final": {}
},
"report_published": "",
"workflow": "Low-pass whole-genome aneuploidy",
"data_links": {
"filtered_variants": "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/Ane_BC18_DeganCases/Ane_BC18_DeganCases_20151014174956618/IR50_Rc10_Ane_BC18_DeganCases_1014_np___4f776c35-4a9f-4a0f-a787-6ee28ebb8c7c.zip",
"unfiltered_variants": "https://baseline.itw/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/Ane_BC18_DeganCases/Ane_BC18_DeganCases_20151014174956618/IR50_Rc10_Ane_BC18_DeganCases_1014_np.zip"
},
"variants_saved": "",
"samples": {
"PROBAND": "Ane_BC18_DeganCases"
},
"shared_with": [],
"stage": "Send for Report Generation",
"flagged": false,
"name": "IR50_Rc10_Ane_BC18_DeganCases_1014_np",
"id": "ff8081815067ca42015068f99193000d",
"ion_reporter_version": "5.0",
"started_by": "Ion User",
"start_date": "October 14, 2015",
"status": "SUCCESSFUL"
}
]
This output contains links to three downloads about this analysis (backslashes have been removed):
-
qc: "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/download/pdf/98ccb759-010f-4c92-acba-2da2d8aa9f4d/ff8081815067ca42015068f99193000d_QC.pdf"
-
unfiltered_variants:" https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/Ane_BC18_DeganCases/Ane_BC18_DeganCases_20151014174956618/IR50_Rc10_Ane_BC18_DeganCases_1014_np.zip"
-
filtered_variants: "https://<server name>/api/v1/download?filepath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/Ane_BC18_DeganCases/Ane_BC18_DeganCases_20151014174956618/IR50_Rc10_Ane_BC18_DeganCases_1014_np___4f776c35-4a9f-4a0f-a787-6ee28ebb8c7c.zip"
curl --location 'https://<servername>/api/v1/analysis?format=json&name=Demo%20AmpliSeq%20CCP%20tumor_prod' \
--header 'Authorization: OTAzNzY1ZjMzY2U4YmE5NzhmN2RhYTFkYTJmMzI2NTE1OGI2YWMzMjRkMGM5OTI1MDkzNWMxMjFiOWRjYWI4MA' \
--header 'Content-Type: application/x-www-form-urlencoded'