CNV Finding parameters
You can adjust Copy Number Variant (CNV) finding parameters to optimize analysis results when you create or edit Ion Reporter™ Software analysis workflows.
IMPORTANT! Use the default parameter settings unless you are an advanced user.
|
Parameter Name |
Description |
|---|---|
|
Main tab |
|
|
Analysis (apply only to Anueploidy analysis workflows) |
|
|
CNV Sensitivity |
Sensitivity. The value of editable parameter Transition Penalty is used by the algorithm only when CUSTOM option is selected, This option is available in Advanced tab for CNV parameters. Allowed values: Low, Medium, High, or Custom |
|
CNV Somatic |
IMPORTANT! The somatic gene-level CNV calling parameter is for beta use only, and requires BED files and a copy number informatics baseline containing gene and pooling information. This parameter is not for use for germline copy number calling such as in ReproSeq workflows or other analysis workflows, which are designed to detect low pass whole genome aneuploidy events. Allowed values: True or False |
|
Advanced tab |
|
|
Gender calling |
|
|
CNV Gender Caller Enable Flag |
Flag to indicate whether Gender caller should be invoked. Allowed values: True or False This flag is valid only for analysis workflows that are used to detect Low Pass whole genome aneuploidy events, such as Ion ReproSeq™ analysis workflows and other low pass whole genome analysis workflows that are used for aneuploidy research. |
|
CNV Gender Threshold |
Specifies threshold ratio of chrY to Autosomes for taking male/female call. Allowed values: 0 to unlimited |
|
CNV Gender Min Mapping QV |
Specifies minimum mapping qv of reads to consider in gender calling. Allowed values: 0 to 255 |
|
CNV Gender Min Autosomes Count |
Specifies minimum number of required filtered reads in autosomes. Allowed values: 0 to unlimited |
|
CNV CHRM to Autosomes Ratio Min Mapping QV |
Specifies minimum mapping qv of reads to consider in calculating chrM A Ratio. Allowed values: 0 to 255 |
|
Sample Filtering (applies only to VCIB CNV finding algorithm) |
|
|
Read Count |
User to enter a threshold number (integer). Allowed values: 10,000 to unlimited |
|
min-mapping-qv |
Minimum mapping quality value required for a read to be counted.
Allowed values: 0 to unlimited |
|
Percent Non Zero Amplicons. Percent of reads aligning to an amplicon in the target regions file. |
User to enter a threshold number (integer). Allowed values: 50 to 100 |
|
MAPD threshold Sample will fail if MAPD is above this threshold. |
User to enter a threshold number (float). Allowed values: 0 to unlimited |
|
Analysis (applies only to VCIB CNV finding algorithm) |
|
|
NPC |
Number of Principal Components used for correction. Allowed values: 0 to 12 |
|
CNV Shift Type |
Allowed values: MEDGENE, MEDAMP, VALUE_BASED_ON_MEDAMP, MAXGENE, or FLATGENE. The following CNV shift methods are available.
|
|
CNV Shift Value |
The amount of subtraction in log2 ratio to the result of the CNV Shift method used. May be used to fix an error in the CNV Shift method. Shift Value is only used when the Shift Type is set to VALUE_BASED_ON_MEDAMP. Allowed values: -2 to 2 |
|
Tumor Cellularity Selection |
Select a value to use tumor calculated tumor fraction or user-specified Percent Tumor Cellularity in sample. Allowed values: Manually input Tumor Cellularity input per user or Auto-calculated tumor cellularity. The minimum amount that cellularity can be adjusted is 40%, for both auto-calculated and manual input tumor cellularity. |
|
Analysis (applies only to aneuploidy analysis workflows) [1] |
|
|
Enable Mosaicism Detection |
Enable Mosaicism Detection Allowed values: True or False |
|
Enable Smoothing |
Enable Smoothing Allowed values: True or False |
|
Set Tile Size for aneuploidy Workflow |
Set the tile size for Ion ReproSeq™ analysis workflows, designed for use with aneuploidy research. The tile size used for creating the aneuploidy baseline must match the tile size selected here. Allowed values: 1 to 10,000,000 |
|
Hide called gender |
Hide gender called by CNV gender calling. Allowed values: True or False |
|
Analysis |
|
|
Plot Y chromosome for Female or Unknown Gender? |
Plot Y chromosome for Female or Unknown Gender. Allowed values: True or False |
|
Analysis (applies only to Exon Deletions) |
|
|
Cutoff for non-integer CN calls |
Specifies the cutoff for making CN #calls that are not precisely of integer values. Allowed values: 0 to 0.50 |
|
Max Calls |
Specifies the number of non-contiguous exon variant calls above which the sample will fail QC. Allowed values: 0 to 47 |
|
Min Quality |
Specifies the quality score below which a CNV variant is classified as a NOCALL. Allowed values: 0 to 100 |
|
Analysis (applies only to Liquid Biopsy and Ion AmpliSeq™ HD) |
|
|
Minimum Tag Family Size |
Minimum number of reads with the same tag required to form a functional family. Allowed values: 0 to unlimited |
|
Max Fold Difference for Loss |
Maximum fold difference relative to reference for calling a loss. Allowed values: 0 to 1 |
|
Min Fold Difference for Gain |
Minimum fold difference relative to reference for calling a gain. Allowed values: 1 to unlimited |
|
P-value for maximum calls |
P value for maximum calls. Allowed values: 0 to 1 |
|
Analysis (applies to all CNV finding algorithm types except VCIB CNV algorithm) |
|
|
CNV Transition Penalty |
Transition Penalty dictates the likelihood that the algorithm will call a different ploidy state between two adjacent data points. Transition Penalty is logarithm (to the base 10) of Transitional Probability. Lower (more negative) values will make it less likely that the algorithm will call adjacent data points as ploidy states that are different from each other. The Transition Penalty parameter edited here will only take effect when using the CUSTOM CNV Sensitivity setting. When CNV MOSAICISM parameter is not enabled, the maximum value supported for Transition Penalty is -1.05. When CNV MOSAICISM parameter is enabled, the maximum value supported for Transition Penalty is -2.31. Allowed values: -1,000,000 to -1.05 |
|
Analysis (applies only to VCIB CNV finding algorithm when custom panel is focal amplification) |
|
|
CNV Gain Threshold |
Threshold value (≥0) for calling GAIN in autosomes Allowed values: 0 to unlimited |
|
CNV Gain Threshold XY |
Threshold value (≥0) for calling GAIN in X or Y for males. Allowed values: 0 to unlimited |
|
Gain Confidence Level |
Confidence level value (>0 and <1) to be used to compare to gain-threshold or gain_threshold_xy. Allowed values: 0 to 1 |
|
Analysis (applies only to Hidden Gender aneuploidy analysis workflow) [2] These settings apply only to aneuploidy workflows which provide only information whether the sex chromosomes look Normal or Abnormal, based on what is expected for the Expected Normal Ploidy of sex chromosomes on a normal XX female or a normal XY male. |
|
|
Male Normal Ploidy Lower Bound |
Male Minimum Normal Ploidy for Hidden Gender aneuploidy analysis workflow. Allowed values: 0 to 100,000 |
|
Male Normal Ploidy Upper Bound |
Male Maximum Normal Ploidy for Hidden Gender aneuploidy analysis workflow Allowed values: 0 to 100,000 |
|
Female Normal Ploidy Lower Bound |
Female Minimum Normal Ploidy for Hidden Gender aneuploidy analysis workflow. Allowed values: 0 to 100,000 |
|
Female Normal Ploidy Upper Bound |
Female Maximum Normal Ploidy for Hidden Gender aneuploidy analysis workflow Allowed values: 0 to 100,000 |