Remove filters
You can remove filters from a filter category from the Workflows table in Ion Reporter Software.
  1. In the Workflows tab, click Overview.
  2. Click the filter category to expand the list, then select Show All at the top of the list.