Sort the Workflows table
You can sort the Workflows table to make it easier to find the analysis workflows that you are looking for in Ion Reporter Software.
  1. In the Workflows tab, in the Overview screen, click a column heading to sort the list of analysis workflows.
  2. Click the column again to reverse the order.
  3. Click the column a third time to return the column to the order that was used before the sort and stop sorting for the column.