HasDeNovoRefAllele
A proband variant is assigned the category HasDeNovoRefAllele if all of the following are true:
-
The variant has an allele that:
-
Is the same as the reference allele.
-
Does not appear in the father's genotype.
-
Does not appear in the mother's genotype.
-
-
The coverage of the father's genotype at this variant position is at least denovo_ref_allele_min_father_coverage.
-
The coverage of the mother's genotype at this variant position is at least denovo_ref_allele_min_mother_coverage.
See also HasDeNovoNonRefAllele.