Genetic Category Type and MAF filter
If a variant is not a minor allele, MAF filter will not be effective, because MAF filter only applies if the variant is a minor allele. Users may have a threshold setting for MAF filter which will not be effective if variant is not a minor allele.