Sort the workflows presets list
You can sort the workflows presets list to make it easier to find the workflow presets that you are looking for in Ion Reporter™ Software.
- In the Workflows tab, click Presets.
- Click a column heading to sort the list based on the column category.
- Click the column heading a third time to return the column to the order that was used before the sort and stop sorting for the column.