Remove filters
You can remove filters from the Version or Reference filter categories in the Workflow Presets table in Ion Reporter™ Software.
- In the Workflows tab, click Presets.
-
Click the Version or Reference filter category to expand the list, then select Show All at the top of the list.
The first filter category does not have a Show All option. You must select a type of workflow preset.