Contigs annotated in the GRCh38 reference genome
Ion Reporter Software annotates only variants that map to the following contigs as provided in the GRCh38.fasta and GRCh38.fasta.fai files.
The following contigs (sequences) are supported by Ion Reporter Software.
Contig Name
Length (bps)
chr1
248956422
chr2
242193529
chr3
198295559
chr4
190214555
chr5
181538259
chr6
170805979
chr7
159345973
chr8
145138636
chr9
138394717
chr10
133797422
chr11
135086622
chr12
133275309
chr13
114364328
chr14
107043718
chr15
101991189
chr16
90338345
chr17
83257441
chr18
80373285
chr19
58617616
chr20
64444167
chr21
46709983
chr22
50818468
chrX
156040895
chrY
57227415
chrM
16569
chr22_KI270879v1_alt
304135
Two kinds of variants are not supported:
  1. Variants that map to a contig we do not support
  2. Variants that map to a masked region
In particular, chr22_KI270879v1_alt is completely masked except for 269,814 - 279,355, which contains only the GSTT1 gene. No amino acid changes are provided for the masked regions. However, gene and transcript information is provided.