Custom panels
The Oncomine™ Variant Annotator plugin applies universal annotation rules for user-defined analysis workflows for use with custom Ion AmpliSeq™ and Ion AmpliSeq™ HD panels from AmpliSeq.com. To apply the rules, the Apply Oncomine Variant Annotator plugin option must be selected when an analysis workflow is created. For more information, see Steps in analysis workflow creation.
IMPORTANT! Because these annotations are generic, the annotations can be different from annotations that are generated with a system-installed analysis workflow.
|
Variant type |
Oncomine™ Gene class |
Oncomine™ Variant class |
Annotation criteria |
|---|---|---|---|
|
Copy number amplification |
Gain-of-Function |
Amplification |
|
|
Copy number deletion |
Loss-of-Function |
Deletion |
|
|
Copy number exon deletion |
Loss-of-Function |
ExonDeletion |
|
|
Copy number exon duplication |
Loss-of-Function |
ExonDuplication |
|
|
Gene fusion |
Gain-of-Function |
Fusion |
|
|
Non-targeted fusion |
Gain-of-Function |
NonTargetedFusion |
|
|
RNA exon variant |
Gain-of-Function |
RNAExonVariant |
|
|
Expression imbalance |
Gain-of-Function |
ExpressionImbalance |
|
|
Long deletion |
Loss-of-Function |
Truncating |
|
|
Loss-of-function truncating de novo mutation |
Loss-of-Function Unclassified |
Truncating |
|
|
Missense hotspot mutation |
Gain-of-Function Loss-of-Function Unclassified |
Hotspot |
|
|
In-frame hotspot mutation |
Gain-of-Function Loss-of-Function Unclassified |
Hotspot |
|
|
Splice site hotspot mutation |
Gain-of-Function Loss-of-Function Unclassified |
Hotspot |
|
|
Intronic hotspot mutation |
Gain-of-Function Loss-of-Function Unclassified |
Hotspot |
|
|
Synonymous hotspot mutation |
Gain-of-Function Loss-of-Function Unclassified |
Hotspot |
|
|
Gain-of-function truncating hotspot mutation |
Gain-of-Function |
Hotspot |
|
|
Loss-of-function truncating hotspot mutation |
Loss-of-Function |
Truncating |
|
|
MNV hotspot mutation |
Gain-of-Function Loss-of-Function Unclassified |
Hotspot |
|
|
CEBPA‑bZIP[1] |
Loss-of-Function |
CEBPA‑bZIP |
|
|
CEBPA‑N‑terminal [1] |
Loss-of-Function |
CEBPA‑N‑terminal |
|
|
EGFR exon 19 deletion |
Gain-of-Function |
EGFRExon19Deletion |
|
|
EGFR exon 20 insertion [1] |
Gain-of-Function |
EGFRExon20Insertion |
|
|
ERBB2 exon 20 insertion [1] |
Gain-of-Function |
ERBB2Exon20Insertion |
|
|
JAK2 exon 12 indel [1] |
Loss-of-Function |
JAK2Exon12Indel |
|
|
KIT exon 11 deletion [1] |
Gain-of-Function |
KITExon11Deletion |
|
|
KIT exon 11 insertion [1] |
Gain-of-Function |
KITExon11Insertion |
|
|
MET exon 14 skipping [1] |
Gain-of-Function |
METExon14Skipping |
|
|
PDGFRA exon 18 deletion [1] |
Gain-of-Function |
PDGFRAExon18Deletion |
|
|
FLT3 internal tandem duplication |
Gain-of-Function |
FLT3ITD |
|